Order Entry
Czech Republic
ContactUsLinkComponent
 
Periodic acid 0.002 mol/l in acetic acid Reag. Ph. Eur. 1063000
Catalog # 87887.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Periodic acid 0.002 mol/l in acetic acid Reag. Ph. Eur. 1063000
Catalog # 87887.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :227.94 g/mol
Storage Temperature:Ambient
EINECS:233-937-0
UN:2920
ADR:8 (3),II
 :GHS02
 :GHS05
CAS Number:10450-60-9
Formula:H₅IO₆
MDL Number:MFCD00011344