Order Entry
Czech Republic
ContactUsLinkComponent
Quartz, BCR® CRM 14-90 μm
Quartz, BCR® CRM 14-90 μm
Catalog # SIALBCR069-10G
Supplier:  Merck
CAS Number:  
undefined
Quartz, BCR® CRM 14-90 μm
Catalog # SIALBCR069-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :60.08 g/mol
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:1427 °C
Density:2.622…2.645 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:238-878-4
CAS Number:14808-60-7
 :7631-86-9
Formula:SiO₂
MDL Number:MFCD00217788