Quartz, BCR® CRM 50-220 μm
Catalog # SIALBCR130-50G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:60.08 g/mol
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:1427 °C
Density:2.622…2.645 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:238-878-4
CAS Number:14808-60-7
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00217788