Order Entry
Czech Republic
ContactUsLinkComponent
Hydrochloric acid 0.25% in methanol Reag. Ph. Eur. 1053203
Flammable
Toxic
Health
Hydrochloric acid 0.25% in methanol Reag. Ph. Eur. 1053203
Catalog # 87868.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Toxic
Health
Hydrochloric acid 0.25% in methanol Reag. Ph. Eur. 1053203
Catalog # 87868.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
ADR:3,II
 :36.46 g/mol
EINECS:231-595-7
UN:1993
 :GHS02
 :GHS06
 :GHS08
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324