Order Entry
Czech Republic
ContactUsLinkComponent
Sodium hydroxide solution 40% (w/w), Biotech Reagent, J.T.Baker®
Corrosive
Irritant
Sodium hydroxide solution 40% (w/w), Biotech Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Irritant
Sodium hydroxide solution 40% (w/w), Biotech Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
    39 - 41%
  • Store above 16°C (60°F) to prevent freezing:
    ≤5 ppm
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤1 ppm
  • Assay (NaOH (in water)):
    39 - 41 %
  • Chloride (Cl):
    ≤25 ppm
  • Heavy Metals (as Pb):
    ≤1 ppm
  • Iron (Fe):
    ≤2 ppm

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together