Sodium chloride 20% in aqueous solution Reag. Ph. Eur. 1079502
Catalog # 87931.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
About this item
Synonyms: Saline, NaCl, Sea salt, Halite
:
:58.44 g/mol
Boiling Pt:1413
Melting Pt:801
Density:1…1.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477
Merck Index:14,08599