Gadolinium trinitrate hexahydrate, Sigma-Aldrich®
Catalog # SIAL217190-10G
Supplier: Merck
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:451.36
Melting Pt:91.5…92 °C
Density:2.4 g/cm³ (20 °C)
EINECS:233-437-2
UN:1477
ADR:5.1,II
CAS Number:19598-90-4
Formula:GdN₃O₉·6H₂O
MDL Number:MFCD00149728