
3,4-Diethoxy-3-cyclobutene-1,2-dione 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5231-87-8
:
Formula:C₈H₁₀O₄
MDL Number:MFCD00001333
:170.16 g/mol
Boiling Pt:95 °C (.1 mmHg)
Density:1.15 g/cm³
Flash Pt:110 °C
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear yellow to light brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5070 to 1.5100 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together