ContactUsLinkComponent
Coomassie brilliant blue G-250, Proteomics Grade
Irritant
Coomassie brilliant blue G-250, Proteomics Grade
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Coomassie brilliant blue G-250, Proteomics Grade
Supplier:  VWR Chemicals
Certificates
Specification Test Results
  • Em (595 nm, Water):
    >36300
  • Protease:
    none detected
  • Protein Staining:
    Pass
  • Appearance:
    Blue to red crystalline powder