Order Entry
Czech Republic
ContactUsLinkComponent
Chromium(III) nitrate nonahydrate, Sigma-Aldrich®
Oxidizing
Irritant
Chromium(III) nitrate nonahydrate, Sigma-Aldrich®
Catalog # SIAL379972-5G
Supplier:  Merck
CAS Number:  
Oxidizing
Irritant
Chromium(III) nitrate nonahydrate, Sigma-Aldrich®
Catalog # SIAL379972-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :400.15 g/mol
Merck Index:13,02246
Melting Pt:60 °C
Density:1.807 g/cm³ (20 °C)
EINECS:236-921-1
UN:2720
ADR:5.1,III
 :M-Clarity™ quality level = MQ100
 :GHS03
 :GHS07
CAS Number:7789-02-8
Formula:CrN₃O₉·9H₂O
MDL Number:MFCD00149659