Order Entry
Czech Republic
ContactUsLinkComponent
Pyridoxine hydrochloride 98+%, extra pure
Pyridoxine hydrochloride 98+%, extra pure
Certificates
undefined
Pyridoxine hydrochloride 98+%, extra pure

Specification Test Results

  • Appearance:
    White to almost white Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.5 % (1 g, 105°C)
  • Sulfated ash:
    ≤0.1 %
  • Solubility:
    (5 % in water) Clear colorless to light yellow
  • pH:
    2.4 to 3.0 (5 % in water)
  • Titration with HClO4:
    98.0 to 102.0 % (On dry basis)
  • Heavy metals (as Pb):
    ≤20 ppm

Specification Test Results

Related Information

Frequently Bought Together