
1-(2-Thenoyl)-3,3,3-trifluoroacetone, (max. 2% H₂O) 99% (dry weight)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:326-91-0
:
Formula:C₈H₅F₃O₂S
MDL Number:MFCD00005445
:222.19 g/mol
Boiling Pt:96…98 °C (8 mmHg)
Melting Pt:41…44 °C
Flash Pt:111 °C (231 °F)
Storage Temperature:Ambient
EINECS:206-316-7
:GHS07
Related Information
Frequently Bought Together