Order Entry
Czech Republic
ContactUsLinkComponent
1-Dodecansulphonic acid sodium salt, LiChropur™ for ion pair chromatography, Supelco®
1-Dodecansulphonic acid sodium salt, LiChropur™ for ion pair chromatography, Supelco®
Catalog # 1.18308.0025
Supplier:  Merck
CAS Number:  
1-Dodecansulphonic acid sodium salt, LiChropur™ for ion pair chromatography, Supelco®
Catalog # 1.18308.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Hill Formula:
    C₁₂H₂₅NaO₃S
  • Molar Mass:
    272.37 g/mol
  • pH value:
    5.5 - 7.5 (100 g/l, H₂O, 20 °C)
  • Bulk density:
    330 kg/m³
  • Solubility:
    (20 °C) soluble
  • Storage class:
    10 - 13 Other liquids and solids
  • WGK:
    WGK 2 water endangering

Specification Test Results

About this item

 :
 :272.38 g/mol
Melting Pt:300 °C
Density:1.218 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:219-200-6
CAS Number:2386-53-0
Formula:C₁₂H₂₅NaO₃S
MDL Number:MFCD00007527