

N,N,3,5-Tetramethylaniline ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4913-13-7
:
Formula:C₁₀H₁₅N
MDL Number:MFCD00008307
:149.24 g/mol
Boiling Pt:226…228 °C
Density:0.913
Flash Pt:90 °C (194 °F)
Storage Temperature:Ambient
EINECS:225-539-0
UN:2810
ADR:6.1,III
:GHS06
:GHS08
Related Information
Frequently Bought Together