


1,2-Diethoxyethane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:629-14-1
:
Formula:C₆H₁₄O₂
MDL Number:MFCD00009253
:118.18 g/mol
Boiling Pt:120…121 °C
Melting Pt:–74 °C
Density:0.842
Flash Pt:19 °C (66 °F)
Storage Temperature:Ambient
EINECS:211-076-1
UN:1153
ADR:3,II
:GHS02
:GHS08
:GHS07
Related Information
Frequently Bought Together