Order Entry
Czech Republic
ContactUsLinkComponent
Chloroform-[D₁] (99.8% D) + 0.03% TMS for NMR spectroscopy
Chloroform-[D₁] (99.8% D) + 0.03% TMS for NMR spectroscopy
  84111.0100
 :  
undefined
Chloroform-[D₁] (99.8% D) + 0.03% TMS for NMR spectroscopy
  84111.0100
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

 

 

  • Assay (on anhydrous substance):
    Min. 99.9 %
  • Isotopic enrichment (FT NMR 400 MHz)(D):
    Min. 99.80 %
  • Water:
    Max. 0.01 %

 

 

 :
 :120.38 g/mol
 :60.8 °C (1013 hPa)
 :–64,1 °C
 :1.5 g/cm³ (20 °C)
 :Refrigerator
 :212-742-4
 :1888
 :6.1,III
 :GHS08
 :GHS07
 :865-49-6
 :CDCl₃
 :MFCD00000827