Human Recombinant Glycoprotein 2 (from HEK293T)
Catalog # ORIGTP307512
Supplier: OriGene
New Product
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:HEK293T cells
- Species:Human
- Tag sequence:C-Myc/DDK
- Storage conditions:Store at –80 °C
- Protein synonyms:ZAP75
- UniProtKB:B7Z1G2
- Protein/peptide name:Glycoprotein 2
- Purity:>80% as determined by SDS-PAGE and Coomassie blue staining
- Molecular weight:42,9 kDa
- Concentration:>0,05 µg/µl as determined by microplate BCA method
Specifications
About this item
This gene encodes an integral membrane protein that is secreted from intracellular zymogen granules and associates with the plasma membrane via glycosylphosphatidylinositol (GPI) linkage. The encoded protein binds pathogens such as enterobacteria, thereby playing an important role in the innate immune response. The C-terminus of this protein is related to the C-terminus of the protein encoded by the neighboring gene, uromodulin (UMOD). Alternative splicing results in multiple transcript variants.