ContactUsLinkComponent
Smooth Muscle Cell Growth Medium
Smooth Muscle Cell Growth Medium
Katalogové číslo PRTV2010601
Dodavatel:  PROVITRO
New Product
Číslo CAS:  
undefined
Smooth Muscle Cell Growth Medium
Katalogové číslo PRTV2010601
Dodavatel:  PROVITRO
New Product
Číslo CAS:  
Specifikace
  • Media Format:
    Liquid
  • Pk:
    500 mL
  • Description:
    Smooth muscle cell growth medium
  • Media Base:
    Basal Medium
Specifikace
O tomto produktu
Smooth Muscle Cell Growth Medium is a sterile liquid culture medium designed for cultivating human smooth muscle cells (HSMC). Provided in the form of a basal medium, it is ideal for culturing HSMC upon the addition of supplement mix components.
  • Formulation is optimised for the initial seeding of 4000 cells/cm², allowing growth up to confluence (approximately 90%)
  • Use of feeder layers, matrix substrates, or additional substances is unnecessary
Due to the possibility of reduced proliferative activity, we recommend to use the antibiotic supplement for freshly isolated cells only. Endothelial cell proliferation medium is thoroughly tested after each production. All components are tested in a stringent biological assay.
Each batch is checked for human endothelial cells proliferating characteristics. The cells cultured in endothelial cell growth medium are checked regarding their morphology, the adherence rate, the colony forming efficiency, and the population doubling time.
Storge: Store the supplemented endothelial cell growth medium in a dark environment at temperatures ranging from 4 to 8 °C for a maximum duration of one month. Avoid heating the medium beyond 37 °C and refrain from using uncontrolled heat sources, such as microwave appliances. When utilising a portion of the medium, extract the required amount from the bottle and warm it appropriately. Never freeze the medium, as this could result in elevated salt concentrations due to the separation of pure water during freezing, leading to irreversible damage.