Pentaerythritol tetrakis [3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate] 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6683-19-8
:
Formula:C₇₃H₁₀₈O₁₂
MDL Number:MFCD00059345
:1177.66 g/mol
Boiling Pt:1006°C (760mmHg)
Storage Temperature:Ambient
EINECS:229-722-6
Specification Test Results
- Appearance:white to almost white powder or crystals
- Infrared spectrum:Conforms
- HPLC:≥94 %
Specification Test Results
Frequently Bought Together