ContactUsLinkComponent
AmberChrom™ chloride form 1x8 200-400 mesh
AmberChrom™ chloride form 1x8 200-400 mesh
Certificates
AmberChrom™ chloride form 1x8 200-400 mesh
Specification Test Results
  • Appearance:
    white to yellow beads
  • Total exchange capacity:
    ≥1.2 eq/l (Cl form)
  • Water retention capacity:
    39 to 45 % (Cl form)
  • Particle size:
    200 to 400 mesh