AmberChrom™ chloride form 1x2 50-100 mesh
:69011-19-4
:MFCD00132718
:(C₁₀H₁₂·C₁₀H₁₀·C₈H₈·C₃H₉N)x
:MFCD00146763
:Ambient
- Appearance:white to yellow beads
- Total exchange capacity:≥0.7 eq/l (Cl form)
- Water retention capacity:65 to 75 % (Cl form)
- Particle size:50 to 100 mesh
Frequently Bought Together