Trihydroxyethylrutin 85%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7085-55-4
:
Formula:C₃₃H₄₂O₁₉
MDL Number:MFCD00893813
:742.7 g/mol
Merck Index:13,09859
Storage Temperature:Refrigerator
EINECS:230-389-4
Specification Test Results
- Appearance:white to yellow to green powder
- Infrared spectrum:Conforms
- HPLC:≥80 %
- Melting point:168°C to 176°C
- Loss on drying:≤5 % (130°C)
Specification Test Results
Frequently Bought Together