Order Entry
Czech Republic
ContactUsLinkComponent
2-(Hydroxymethyl)-1,3-propanediol 98%
Irritant
2-(Hydroxymethyl)-1,3-propanediol 98%
Catalog # 470770010.
CAS Number:  
Irritant
2-(Hydroxymethyl)-1,3-propanediol 98%
Catalog # 470770010.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    white powder or crystals or chunks
  • Infrared spectrum:
    Conforms
  • GC:
    ≥97.5 %
  • Melting point:
    63°C to 68°C

Specification Test Results

About this item

 :
 :106.12 g/mol
EINECS:225-187-8
 :GHS07
Storage Temperature:Ambient
CAS Number:4704-94-3
Formula:C₄H₁₀O₃
MDL Number:MFCD00192217