
Naratriptan hydrochloride 95%
Catalog # 470752500.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mg
- Seq:2590
Specifications
Specification Test Results
- Appearance:white to beige powder
- Infrared spectrum:Conforms
- HPLC:≥94 %
- Melting point:234°C to 246°C
Specification Test Results
About this item
:
:371.92 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:143388-64-1
Formula:C₁₇H₂₅N₃O₂S·HCl
MDL Number:MFCD08141821