

4-[(Methylsulphonyl)amino]benzoic acid 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7151-76-0
:
Formula:C₈H₉NO₄S
MDL Number:MFCD00025052
:215.226 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:white to yellow powder
- Infrared spectrum:Conforms
- HPLC:≥94 %
- Titration with NaOH:94 to 106 %
Specification Test Results
Frequently Bought Together