
2-(1-Pyrazolyl)pyridine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25700-11-2
:
Formula:C₈H₇N₃
MDL Number:MFCD09033534
:145.17 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:white to almost white crystalline powder and or chunks
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Melting point:35°C to 45°C
Specification Test Results
Frequently Bought Together