Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Přihlásit se
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pigment Red 22
Pigment Red 22
Pigment Red 22
Pigment Red 22
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Pigment Red 22
Supplier:
Thermo Fisher Scientific
About this item
Specification Test Results
Appearance:
red powder
Infrared spectrum:
Conforms
Loss on drying:
≤2 %
Specification Test Results
Frequently Bought Together
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.5% (by anhydrous basis) stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol, LiChrosolv®, hypergrade for LC-MS, for HPLC, Supelco®
Supplier: Merck
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Tetrahydrofuran, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
tert-Butyl methyl ether, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethyl acetate, anhydrous (max. 0.005% H₂O) ≥99.8%
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals