
4'-Trifluoromethylbiphenyl-4-carbaldehyde
:90035-34-0
:
:C₁₄H₉F₃O
:MFCD01862519
:250.2200 g/mol
:GHS07
:Ambient
- Appearance:white to light yellow powder or crystals
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Melting point:74°C to 78°C
Frequently Bought Together