
2,6-Dibromo-9,10-anthraquinone
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:633-70-5
:
Formula:C₁₄H₆Br₂O₂
MDL Number:MFCD08276344
:366.0042 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:light yellow to brown crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥94 %
Specification Test Results
Frequently Bought Together