


2,2-Dibromo-2-cyanoacetamide 90%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10222-01-2
:
Formula:C₃H₂Br₂N₂O
MDL Number:MFCD00129791
:241.87 g/mol
Boiling Pt:221°C (760mmHg)
Storage Temperature:Ambient
EINECS:233-539-7
UN:2811
ADR:6.1,II
:GHS05
:GHS06
:GHS09
Specification Test Results
- Appearance:white to light yellow crystalline powder
- Infrared spectrum:Conforms
- GC:≥88 %
- Melting point:122°C to 127°C
Specification Test Results
Frequently Bought Together