
1,6-Hexanediol dimethacrylate 95% stabilised
:6606-59-3
:
:C₁₄H₂₂O₄
:MFCD00015425
:254.33 g/mol
:>315°C
:0.995
:109°C
:229-551-7
:GHS07
:Ambient
- Appearance:clear colorless liquid
- Infrared spectrum:Conforms
- GC:≥94 %
- Refractive index:1.4570 to 1.4590 (20°C, 589 nm)
- Stabilizer:ca. 200 ppm MeHQ
Frequently Bought Together