ContactUsLinkComponent
Silver nitrate 99+%, meets analytical specification of Ph.Eur.
Oxidizing
Corrosive
Health
Environment
Silver nitrate 99+%, meets analytical specification of Ph.Eur.
Certificates
Oxidizing
Corrosive
Health
Environment
Silver nitrate 99+%, meets analytical specification of Ph.Eur.
Specification Test Results
  • Appearance:
    white to almost white transparant colorless crystalline powder or crystals
  • Titration with NH4SCN:
    99.0 to 100.5 %
  • Acidity/alkalinity:
    Passes test
  • Appearance of solution:
    (2 g in 50 ml water) Clear and colorless