2-Fluoroethylamine hydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:460-08-2
:
Formula:C₂H₆FN·HCl
MDL Number:MFCD00008161
:99.53 g/mol
Storage Temperature:Ambient
EINECS:207-302-3
Specification Test Results
- Appearance:white to light yellow to light orange crystalline powder
- Infrared spectrum:Conforms
- Titration Argentometric:96.0 to 104.0 %
- Melting point:99°C to 103°C
- Water:≤4 %
Specification Test Results
Frequently Bought Together