
1,2,3,4-Tetrahydro-2-naphthoic acid 98%
:53440-12-3
:
:C₁₁H₁₂O₂
:MFCD00001741
:176.212 g/mol
:343.9°C
:258-553-0
:GHS07
:Ambient
- Appearance:white to yellow powder or crystals
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Melting point:93°C to 96°C
- Titration with NaOH:97.5 to 102.5 %
Frequently Bought Together