Dibenzothiophene-5,5-dioxide 98%
:1016-05-3
:
:C₁₂H₈O₂S
:MFCD00004970
:216.25 g/mol
:Ambient
:213-805-9
- Appearance:white to yellow crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Melting point:231°C to 236°C
Frequently Bought Together