Order Entry
Czech Republic
ContactUsLinkComponent
Methyl green 0.1% (w/v) in aqueous solution
Methyl green 0.1% (w/v) in aqueous solution
Catalog # 42747.AE
CAS Number:  
Methyl green 0.1% (w/v) in aqueous solution
Catalog # 42747.AE
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :516.95 g/mol
Merck Index:14,6082
Storage Temperature:Ambient
EINECS:238-920-1
CAS Number:14855-76-6
Formula:C₂₇H₃₅BrClN₃
MDL Number:MFCD00036449