Order Entry
Czech Republic
ContactUsLinkComponent
Hydrochloric acid, Test Solution (TS)
Hydrochloric acid, Test Solution (TS)
Catalog # ACSDCH8002068.1L
Supplier:  CPACHEM
New Product
CAS Number:  
Hydrochloric acid, Test Solution (TS)
Catalog # ACSDCH8002068.1L
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :36.46 g/mol
Storage Temperature:Room Temperature
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324