Order Entry
Czech Republic
ContactUsLinkComponent
Potassium sulphate standard solution 100 µg/ml SO₄
Potassium sulphate standard solution 100 µg/ml SO₄
Catalog # ACSDCH0802.L1
Supplier:  CPACHEM
New Product
CAS Number:  
Potassium sulphate standard solution 100 µg/ml SO₄
Catalog # ACSDCH0802.L1
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :174.26 g/mol
Storage Temperature:Room Temperature
CAS Number:7778-80-5
Formula:K₂SO₄
MDL Number:MFCD00011388