Chlorid sodný standard solution (stock)
Catalog # ACSDCH0801.L1
Supplier: CPACHEM
New Product
CAS Number:
Specifications
- Bal.:100 mL
- Sekvence:2440
Specifications
About this item
:
Storage Temperature:Room Temperature
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477