Order Entry
Czech Republic
ContactUsLinkComponent
Nickel Standard Solution, 1 g/L Ni in dil. Nitric Acid for AAS
Nickel Standard Solution, 1 g/L Ni in dil. Nitric Acid for AAS
Katalogové číslo ACSDA038.2NP.L1
Dodavatel:  CPACHEM
New Product
Číslo CAS:  
Nickel Standard Solution, 1 g/L Ni in dil. Nitric Acid for AAS
Katalogové číslo ACSDA038.2NP.L1
Dodavatel:  CPACHEM
New Product
Číslo CAS:  

Specifikace

  • Pk:
    100 mL
  • Description:
    Nickel standard solution
  • Storage conditions:
    Store under normal laboratory conditions.
  • Hazard class description:
    Corrosive substances
  • UN No.:
    1760
  • GHS signal:
    Danger
  • Storage temperature:
    15...25 °C
  • Physical form:
    Fluid
  • Element:
    Nickel
  • Symbol:
    Ni

Specifikace

O tomto produktu

Ordering information: Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.