2'-Deoxy-3'-O-(N'-methylanthraniloyl)adenosine-5'-O-diphosphate sodium salt ≥95%
Catalog # CAYM38670-2.7
Supplier: Cayman Chemical
New Product
CAS Number:
Specifications
- Pk:2,7 mg
- Seq:2175
Specifications
About this item
:
:610.3 g/mol
Boiling Pt:100 °C (212 °F)
Melting Pt:0 °C (32 °F)
Storage Temperature:Freezer
:91828-83-0
Formula:C₁₈H₁₉N₆O₁₀P₂·3Na