Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
La-Pha-Pack® Snap Ring Cap

La-Pha-Pack® Snap Ring Cap

Snap ring cap soft version (transparent), 45 mm, PTFE
Catalog # LAPH11151983
Supplier:   LA PHA PACK
La-Pha-Pack® Snap Ring Cap
Catalog # LAPH11151983
Supplier:  LA PHA PACK
La-Pha-Pack® Snap Ring Cap

Snap ring cap soft version (transparent), 45 mm, PTFE

Price
7 310,00 Kč
Log in for availability
1 * 1.000 KS
Product Details & Documents

Snap ring cap soft version (transparent) with septa red rubber/PTFE (red/beige), 1,0 mm.

Specifications
  • Ø:
    45 mm
  • Description:
    Snap ring cap soft version (transparent)
  • Material:
    PTFE
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.