Ammonium thiocyanate 0,1 M according to (ChP)
Catalog # REGECPAT2010F
Supplier: REAGECON
New Product
CAS Number:
Specifications
- Pack type:Amber glass Winchester bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:76.12 g/mol
CAS Number:1762-95-4
Formula:NH₄SCN
MDL Number:MFCD00011428