Order Entry
Czech Republic
ContactUsLinkComponent
Barium chloride 0,1 M ChP
Irritant
Barium chloride 0,1 M ChP
Catalog # REGECPBACL2101
Supplier:  REAGECON
New Product
CAS Number:  
Irritant
Barium chloride 0,1 M ChP
Catalog # REGECPBACL2101
Supplier:  REAGECON
New Product
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle GL45
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :208.24 g/mol
 :GHS07
CAS Number:10361-37-2
Formula:BaCl₂
MDL Number:MFCD00003445