
Magnesium ≥99.0%, ribbon, GPR RECTAPUR®
Catalog # 25071.132
Supplier: VWR Chemicals
New Product
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Assay (Magnesium):min. 99.0%
- Iron (Fe):max. 0.01%
- Lead (Pb):max. 0.01%
- Manganese (Mn):max. 0.030%
- Nickel (Ni):max. 0.005%
- Zinc (Zn):max. 0.01%
Specification Test Results
About this item
:
:24.3 g/mol
Boiling Pt:1107 °C (1013.25 hPa)
Melting Pt:651 °C
Density:1.75 g/cm3 (20 °C)
Storage Temperature:Ambient
EINECS:231-104-6
UN:1869
ADR:4.1,III
:GHS02
CAS Number:7439-95-4
Formula:Mg
MDL Number:MFCD00085308