Order Entry
Czech Republic
ContactUsLinkComponent
Potassium pyroantimonate 0.05 mol/l in potassium hydroxide solution Reag. Ph. Eur. 1071301
Corrosive
Environment
Potassium pyroantimonate 0.05 mol/l in potassium hydroxide solution Reag. Ph. Eur. 1071301
Catalog # 87725.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Environment
Potassium pyroantimonate 0.05 mol/l in potassium hydroxide solution Reag. Ph. Eur. 1071301
Catalog # 87725.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :262.9 g/mol
EINECS:235-387-7
UN:3266
ADR:8,III
 :GHS05
 :GHS09
CAS Number:12208-13-8
 :MFCD00150439
Formula:KSbO₃·3H₂O
MDL Number:MFCD00011415