VWR®, Endosulfan mixed isomers neat Certified Reference Material
Catalog # 124122S
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass ampoule
- Pk:10 mg
- CAS No.:115-29-7
- Description:Endosulfan, mixed isomers neat CRM
- Storage conditions:Ambient (>5 °C)
- Formula:C₉H₆Cl₆O₃S
- Molecular weight:406.93 g/mol
- Description:Endosulfan, mixed isomers
Specifications
About this item
- Purity (GC/MS) 99.5 %