

Formaldehyde 4% for histology, buffered pH 6.9, Sigma-Aldrich®
Catalog # 1.00496.5000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic can
- Pk:5 L
- Seq:5190
Specifications
Specification Test Results
- Boiling Point:100 °C (1013 hPa)
- Density:1.017 g/cm3 (20 °C)
- pH:6.7 - 7.1 (H2O)
Specification Test Results
About this item
:
:30.03 g/mol
:M-Clarity™ quality level = MQ400
EINECS:200-001-8
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274