Přeskočit na Záhlaví
Přeskočit na Hlavní obsah
Přeskočit na Patička
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Magnesium silicate
Florisil® for chromatography
Florisil® for chromatography
Florisil® for chromatography
PROOSC-4183-B005
:
LGC Standards PROMOCHEM
:
Florisil® for chromatography
PROOSC-4183-B005
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle for solids
Pk:
500 g
Seq:
3645
:
:
100.38 g/mol
:
215-681-1
:
1343-88-0
:
MgO₃Si
:
MFCD00078249