Order Entry
Czech Republic
ContactUsLinkComponent
 
di-Ammonium oxalate solution Reag. Ph. Eur. 1005901
Catalog # 87774.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
di-Ammonium oxalate solution Reag. Ph. Eur. 1005901
Catalog # 87774.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :142.11 g/mol
Storage Temperature:Ambient
EINECS:214-202-3
CAS Number:6009-70-7
Formula:(NH₄)₂C₂O₄·H₂O
MDL Number:MFCD00149694